Assay Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040581.2 | NP_001035671.1 | 29 | 55 | ||
| GenBank mRNA | BC108180.1 | - | 30 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108180.1, NM_001040581.2 transcripts.
Set Membership: |
|

RPS21
Genomic Map