Assay Details
| Gene Symbol: CDKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040582.2 | NP_001035672.1 | 5 - 6 | 322 | 84 | |
| GenBank mRNA | BC108173.1 | - | 5 - 6 | 324 | 84 | |
More Information
Set Membership: |
|

CDKN3
Genomic Map