Assay Details
| Gene Symbol: MUSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040589.2 | NP_001035679.1 | 2 - 3 | 200 | 82 | |
| GenBank mRNA | BC109919.1 | - | 2 - 3 | 201 | 82 | |
More Information
Set Membership: |
|

MUSTN1
Genomic Map