Assay Details
| Gene Symbol: RHOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040602.2 | NP_001035692.1 | 2 - 3 | 252 | 61 | |
| GenBank mRNA | BC103016.1 | - | 2 - 3 | 253 | 61 | |
More Information
Set Membership: |
|

RHOF
Genomic Map