Assay Details
| Gene Symbol: LOC505033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045907.2 | NP_001039372.1 | 1 - 2 | 332 | 92 | |
| GenBank mRNA | BC114179.1 | - | 1 - 2 | 333 | 92 | |
More Information
Set Membership: |
|

LOC505033
Genomic Map