Assay Details
| Gene Symbol: LOC505033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045907.2 | NP_001039372.1 | 2 - 3 | 448 | 81 | |
| GenBank mRNA | BC114179.1 | - | 2 - 3 | 449 | 81 | |
More Information
Set Membership: |
|

LOC505033
Genomic Map