Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045951.1 | NP_001039416.1 | 2 - 3 | 216 | 71 | |
| GenBank mRNA | BC111637.1 | - | 2 - 3 | 216 | 71 | |
More Information
Set Membership: |
|

CARHSP1
Genomic Map