Assay Details
| Gene Symbol: CHCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046484.2 | NP_001039949.1 | 1 - 2 | 128 | 93 | |
| GenBank mRNA | BC105502.1 | - | 1 - 2 | 129 | 93 | |
More Information
Set Membership: |
|

CHCHD1
Genomic Map