Assay Details
| Gene Symbol: SRPRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075122.1 | NP_001068590.1 | 8 - 9 | 1111 | 66 | |
| GenBank mRNA | BC105264.1 | - | 8 - 9 | 1111 | 66 | |
More Information
Set Membership: |
|

SRPRA
Genomic Map