Assay Details
| Gene Symbol: ATP5F1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075136.2 | NP_001068604.1 | 3 - 4 | 252 | 92 | |
| GenBank mRNA | BC102466.1 | - | 3 - 4 | 253 | 92 | |
More Information
Set Membership: |
|

ATP5F1C
Genomic Map