Assay Details
| Gene Symbol: PPP1R42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075238.3 | NP_001068706.2 | 5 - 6 | 809 | 73 | |
| NR_153377.1 | 3 - 4 | 439 | 73 | |||
| GenBank mRNA | BC102655.1 | - | 6 - 7 | 766 | 73 | |
| DT855618.1 | - | 5 - 6 | 767 | 73 | ||
| DT860989.1 | - | 2 - 3 | 160 | 73 | ||
| EH202590.1 | - | 3 - 4 | 366 | 73 | ||
More Information
Set Membership: |
|

PPP1R42
Genomic Map