Assay Details
| Gene Symbol: CHP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075576.1 | NP_001069044.1 | 7 - 8 | 798 | 73 | |
| GenBank mRNA | BC103418.1 | - | 7 - 8 | 798 | 73 | |
| BT030715.1 | - | 6 - 7 | 756 | 73 | ||
More Information
Set Membership: |
|

CHP1
Genomic Map