Assay Details
| Gene Symbol: FABP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075675.2 | NP_001069143.1 | 3 - 4 | 385 | 94 | |
| GenBank mRNA | BC102199.1 | - | 3 - 4 | 386 | 94 | |
More Information
Set Membership: |
|

FABP6
Genomic Map