Assay Details
| Gene Symbol: CMTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075807.1 | NP_001069275.1 | 2 - 3 | 522 | 64 | |
| GenBank mRNA | BC109647.1 | - | 2 - 3 | 522 | 64 | |
More Information
Set Membership: |
|

CMTM2
Genomic Map