Assay Details
| Gene Symbol: SCAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076054.2 | NP_001069522.1 | 4 - 5 | 397 | 63 | |
| GenBank mRNA | BC102448.1 | - | 4 - 5 | 398 | 63 | |
| BT026177.1 | - | 4 - 5 | 375 | 63 | ||
More Information
Set Membership: |
|

SCAMP1
Genomic Map