Assay Details
| Gene Symbol: CD164L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076180.1 | NP_001069648.1 | 2 - 3 | 275 | 71 | |
| GenBank mRNA | BC108248.1 | - | 2 - 3 | 275 | 71 | |
More Information
Set Membership: |
|

CD164L2
Genomic Map