Assay Details
| Gene Symbol: FKBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076391.4 | NP_001069859.2 | 1 - 2 | 377 | 120 | |
| GenBank mRNA | BC107539.1 | - | 1 - 2 | 378 | 120 | |
More Information
Set Membership: |
|

FKBP14
Genomic Map