Assay Details
| Gene Symbol: SPA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076440.2 | NP_001069908.1 | 4 - 5 | 388 | 136 | |
| GenBank mRNA | BC103421.1 | - | 4 - 5 | 390 | 136 | |
More Information
Set Membership: |
|

SPA17
Genomic Map