Assay Details
| Gene Symbol: POLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076481.2 | NP_001069949.1 | 2 - 3 | 378 | 58 | |
| GenBank mRNA | BC102212.1 | - | 2 - 3 | 379 | 58 | |
More Information
Set Membership: |
|

POLD4
Genomic Map