Assay Details
| Gene Symbol: POLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076481.2 | NP_001069949.1 | 3 - 4 | 487 | 111 | |
| GenBank mRNA | BC102212.1 | - | 3 - 4 | 488 | 111 | |
More Information
Set Membership: |
|

POLD4
Genomic Map