Assay Details
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076516.2 | NP_001069984.1 | 2 - 3 | 335 | 96 | |
| GenBank mRNA | BC108196.1 | - | 2 - 3 | 337 | 96 | |
More Information
Set Membership: |
|

PHPT1
Genomic Map