Assay Details
| Gene Symbol: FMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076997.1 | NP_001070465.1 | 1 - 2 | 145 | 62 | |
| GenBank mRNA | BC103015.1 | - | 1 - 2 | 145 | 62 | |
More Information
Set Membership: |
|

FMC1
Genomic Map