Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077005.2 | NP_001070473.1 | 7 - 8 | 1170 | 61 | |
| GenBank mRNA | BC103353.1 | - | 7 - 8 | 1171 | 61 | |
More Information
Set Membership: |
|

AZI2
Genomic Map