Assay Details
| Gene Symbol: DMAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077007.2 | NP_001070475.1 | 2 - 3 | 363 | 104 | |
| GenBank mRNA | BC103419.1 | - | 2 - 3 | 364 | 104 | |
More Information
Set Membership: |
|

DMAC1
Genomic Map