Assay Details
| Gene Symbol: CXHXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077032.2 | NP_001070500.1 | 1 - 2 | 205 | 93 | |
| GenBank mRNA | BC109548.1 | - | 1 - 2 | 206 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109548.1, NM_001077032.2 transcripts.
Set Membership: |
|

CXHXorf65
Genomic Map