Assay Details
| Gene Symbol: CXHXorf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077032.2 | NP_001070500.1 | 4 - 5 | 502 | 65 | |
| GenBank mRNA | BC109548.1 | - | 4 - 5 | 503 | 65 | |
More Information
Set Membership: |
|

CXHXorf65
Genomic Map