Assay Details
| Gene Symbol: SDHAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077045.2 | NP_001070513.1 | 2 - 3 | 350 | 126 | |
| GenBank mRNA | BC103446.1 | - | 2 - 3 | 351 | 126 | |
More Information
Set Membership: |
|

SDHAF2
Genomic Map