Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077119.2 | NP_001070587.1 | 3 - 4 | 303 | 124 | |
| GenBank mRNA | BC108217.1 | - | 3 - 4 | 304 | 124 | |
More Information
Set Membership: |
|

DYNLRB2
Genomic Map