Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077830.2 | NP_001071298.1 | 3 - 4 | 354 | 84 | |
| GenBank mRNA | AB099089.1 | - | 3 - 4 | 334 | 84 | |
| BC102536.1 | - | 3 - 4 | 355 | 84 | ||
More Information
Set Membership: |
|

RPS14
Genomic Map