Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077830.2 | NP_001071298.1 | 4 - 5 | 429 | 61 | |
| GenBank mRNA | BC102536.1 | - | 4 - 5 | 430 | 61 | |
More Information
Set Membership: |
|

RPS14
Genomic Map