Assay Details
| Gene Symbol: SLC9A3R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077852.2 | NP_001071320.1 | 4 - 5 | 1053 | 62 | |
| GenBank mRNA | BC102807.1 | - | 4 - 5 | 1054 | 62 | |
More Information
Set Membership: |
|

SLC9A3R1
Genomic Map