Assay Details
| Gene Symbol: PRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078053.2 | NP_001071521.1 | 1 | 351 | 95 | |
| GenBank mRNA | BC108198.1 | - | 1 | 352 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRM3| Gene Symbol: PRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078053.2 | NP_001071521.1 | 1 | 351 | 95 | |
| GenBank mRNA | BC108198.1 | - | 1 | 352 | 95 | |
Set Membership: |
|