Assay Details
| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078059.2 | NP_001071527.2 | 10 - 11 | 1114 | 86 | |
| GenBank mRNA | BC105169.1 | - | 10 - 11 | 1114 | 86 | |
More Information
Set Membership: |
|

C2CD6
Genomic Map