Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078127.2 | NP_001071595.1 | 1 - 2 | 58 | 144 | |
| GenBank mRNA | BC102840.1 | - | 1 - 2 | 59 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102840.1, NM_001078127.2 transcripts.
Set Membership: |
|

YWHAQ
Genomic Map