Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078127.2 | NP_001071595.1 | 5 - 6 | 814 | 68 | |
| GenBank mRNA | BC102840.1 | - | 5 - 6 | 815 | 68 | |
| BC151414.1 | - | 4 - 5 | 736 | 68 | ||
| BT030749.2 | - | 4 - 5 | 768 | 68 | ||
More Information
Set Membership: |
|

YWHAQ
Genomic Map