Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078132.2 | NP_001071600.1 | 1 | 29 | 56 | |
| GenBank mRNA | BC102179.1 | - | 1 | 30 | 56 | |
| BC148017.1 | - | 1 - 2 | 31 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102179.1, BC148017.1, NM_001078132.2 transcripts.
Set Membership: |
|

RPL37
Genomic Map