Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078132.2 | NP_001071600.1 | 1 | 170 | 71 | |
| GenBank mRNA | BC102179.1 | - | 1 | 171 | 71 | |
| BC148017.1 | - | 2 - 3 | 172 | 71 | ||
More Information
Set Membership: |
|

RPL37
Genomic Map