Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372082.1 | NP_001359011.1 | 1 - 2 | 347 | 110 | |
| GenBank mRNA | BC104529.1 | - | 1 - 2 | 341 | 110 | |
More Information
Set Membership: |
|

NUAK2
Genomic Map