Assay Details
| Gene Symbol: TMEM80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080312.2 | NP_001073781.1 | 4 - 5 | 231 | 63 | |
| GenBank mRNA | BC126790.1 | - | 4 - 5 | 232 | 63 | |
More Information
Set Membership: |
|

TMEM80
Genomic Map