Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080313.2 | NP_001073782.1 | 4 - 5 | 344 | 69 | |
| GenBank mRNA | BC122780.1 | - | 4 - 5 | 345 | 69 | |
More Information
Set Membership: |
|

WASHC3
Genomic Map