Assay Details
| Gene Symbol: ASS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173892.4 | NP_776317.1 | 2 - 3 | 151 | 76 | |
| GenBank mRNA | BC102474.1 | - | 2 - 3 | 152 | 76 | |
| M26198.1 | - | 1 - 2 | 117 | 76 | ||
More Information
Set Membership: |
|

ASS1
Genomic Map