Assay Details
| Gene Symbol: ASS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173892.4 | NP_776317.1 | 3 - 4 | 222 | 67 | |
| GenBank mRNA | BC102474.1 | - | 3 - 4 | 223 | 67 | |
| M26198.1 | - | 2 - 3 | 188 | 67 | ||
More Information
Set Membership: |
|

ASS1
Genomic Map