Assay Details
| Gene Symbol: ASS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173892.4 | NP_776317.1 | 5 - 6 | 464 | 67 | |
| GenBank mRNA | BC102474.1 | - | 5 - 6 | 465 | 67 | |
| M26198.1 | - | 4 - 5 | 430 | 67 | ||
More Information
Set Membership: |
|

ASS1
Genomic Map