Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173929.3 | NP_776354.2 | 3 - 4 | 349 | 63 | |
| GenBank mRNA | BC108213.1 | - | 3 - 4 | 349 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAEP| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173929.3 | NP_776354.2 | 3 - 4 | 349 | 63 | |
| GenBank mRNA | BC108213.1 | - | 3 - 4 | 349 | 63 | |
Set Membership: |
|