Assay Details
| Gene Symbol: TBCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175803.3 | NP_786997.1 | 1 - 2 | 132 | 133 | |
| GenBank mRNA | BC109909.1 | - | 1 - 2 | 132 | 133 | |
More Information
Set Membership: |
|

TBCA
Genomic Map