Assay Details
| Gene Symbol: TBCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175803.3 | NP_786997.1 | 2 - 3 | 239 | 85 | |
| GenBank mRNA | BC109909.1 | - | 2 - 3 | 239 | 85 | |
| X97224.1 | - | 163 | 85 | |||
More Information
Set Membership: |
|

TBCA
Genomic Map