Assay Details
| Gene Symbol: CAPZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176648.4 | NP_788821.1 | 4 - 5 | 485 | 63 | |
| GenBank mRNA | BC102613.1 | - | 4 - 5 | 486 | 63 | |
| Y10372.1 | - | 424 | 63 | |||
| Z85980.1 | - | 424 | 63 | |||
More Information
Set Membership: |
|

CAPZB
Genomic Map