Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205801.2 | NP_991370.1 | 1 | 755 | 117 | |
| GenBank mRNA | AB115781.1 | - | 1 | 722 | 117 | |
| BC102398.1 | - | 1 | 755 | 117 | ||
More Information
Set Membership: |
|

CLDN3
Genomic Map