Assay Details
| Gene Symbol: CRELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014851.3 | NP_001014851.3 | 7 - 8 | 813 | 54 | |
| GenBank mRNA | BC109768.1 | - | 7 - 8 | 770 | 54 | |
| BT020723.1 | - | 7 - 8 | 769 | 54 | ||
| BT021876.1 | - | 6 - 7 | 753 | 54 | ||
More Information
Set Membership: |
|

CRELD1
Genomic Map