Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025321.3 | NP_001020492.1 | 372 | 97 | ||
| GenBank mRNA | BC111654.1 | - | 373 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL35A| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025321.3 | NP_001020492.1 | 372 | 97 | ||
| GenBank mRNA | BC111654.1 | - | 373 | 97 | ||
Set Membership: |
|